Skip to Content
Merck
All Photos(2)

Key Documents

900029

Sigma-Aldrich

Polyphthalaldehyde

Linear

Synonym(s):

PPA

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$2,186.00

R$2,186.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$2,186.00

About This Item

Linear Formula:
(C8H6O2)n
UNSPSC Code:
12352200
NACRES:
NA.23

R$2,186.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

mol wt

average Mn 5000-8000

PDI

<2

Looking for similar products? Visit Product Comparison Guide

Application

PPA is a self-amplified depolymerization media for dry lithography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probe-based 3-D nanolithography using self-amplified depolymerization polymers.
Armin W Knoll et al.
Advanced materials (Deerfield Beach, Fla.), 22(31), 3361-3365 (2010-04-27)
Wanji Seo et al.
Journal of the American Chemical Society, 132(27), 9234-9235 (2010-06-23)
This Communication describes a strategy for designing stimuli-responsive plastics that are capable of responding to chemical signals in the environment by changing shape. The plastics consist of patterned mixtures of poly(phthalaldehyde) polymers in which each polymer contains a different end-capping

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service