Skip to Content
Merck
All Photos(1)

Key Documents

89170

Sigma-Aldrich

Thorium oxide

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ThO2
CAS Number:
Molecular Weight:
264.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.0%

reaction suitability

reagent type: catalyst
core: thorium

extent of labeling

7.15 kBq/g specific activity

SMILES string

O=[Th]=O

InChI

1S/2O.Th

InChI key

ZCUFMDLYAMJYST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

Regulations require a license for the sale of radioactive substances in Switzerland. Swiss customers are kindly requested when ordering, to include their license number, issued by the Swiss Public Health Authorities.

Other Notes

Sales restrictions may apply
Regulations require a license for the sale of radioactive substances in Switzerland. Swiss customers are kindly requested when ordering, to include their license number, issued by the Swiss Public Health Authorities.

Pictograms

Health hazardSkull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Carcinogenic chemicals and risk of hematopoietic neoplasms].
Masanobu Kobayashi
Nihon rinsho. Japanese journal of clinical medicine, 70 Suppl 2, 39-42 (2012-11-09)
Yoichiro Yamamoto et al.
Health physics, 99(3), 388-393 (2010-08-12)
Irradiation from internally deposited radionuclides induces malignant tumors. Ingested radionuclides accumulate in specific organs, which are irradiated over a lifelong period. Our aim is to elucidate why the development of malignant tumors requires long-term internal exposure, on the order of
Rakesh K Behera et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(21), 215405-215405 (2012-05-12)
Thorium dioxide (ThO(2)) is of great interest to energy research as thorium-based nuclear fuel offers the promise of increased proliferation resistance, longer fuel cycles, higher burn-up and improved wasteform characteristics in the generation of nuclear power. However, understanding of ThO(2)
G van Kaick et al.
Der Radiologe, 51(12), 1052-1057 (2011-11-18)
The medical management and counseling of persons at high risk due to exposure to chemicals or radiation or due to personal disposition, present an additional challenge for physicians and especially radiologists involved. This article is based on own experiences with
Thorotrast toxicity: the safety of gadolinium compounds.
J F M Wetzels
The Netherlands journal of medicine, 65(8), 276-278 (2007-09-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service