Skip to Content
Merck
All Photos(1)

Key Documents

89095

Sigma-Aldrich

(R)-Tetrahydrofurfuryl alcohol

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(R)-Tetrahydrofuran-2-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
Beilstein:
79846
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical purity

enantiomeric ratio: ≥97:3% (GC)

bp

178 °C (lit.)

density

1.054 g/mL at 20 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OC[C@H]1CCCO1

InChI

1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2/t5-/m1/s1

InChI key

BSYVTEYKTMYBMK-RXMQYKEDSA-N

Application

(R)-Tetrahydrofurfuryl alcohol can be used:
  • To prepare (R)-tetrahydrofurfuryl aldehyde, which is employed as a building block to synthesize a fragment of pectenotoxin-4.[1]
  • As a starting material to synthesize naftidrofuryl isomers, used as potent 5-hydroxytryptamine 2A receptor antagonists.[2]
  • As a building block for the preparation of (S)-2-tetrahydrofuran methyl ether 6-fluorophenyl ether analog, as an IGF-1R inhibitor.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-catalysed vinyl 1, 4-conjugate addition coupled with Sharpless asymmetric dihydroxylation in the synthesis of the CDE ring fragment of pectenotoxin-4
Richardson MSW, et al.
Chemical Science, 10(25), 6336-6340 (2019)
Optimisation of a 5-[3-phenyl-(2-cyclic-ether)-methyl-ether]-4-aminopyrrolopyrimidine series of IGF-1R inhibitors
Fairhurst RA, et al.
Bioorganic & medicinal chemistry letters, 26(8), 2057-2064 (2016)
Practical access to four stereoisomers of naftidrofuryl and their binding affinity towards 5-hydroxytryptamine 2A receptor
Hao J, et al.
Bioorganic & Medicinal Chemistry Letters, 22(10), 3441-3444 (2012)
[Acute poisoning with tetrahydrofurfuryl alcohol combined with methanol].
B F Murashov et al.
Voenno-meditsinskii zhurnal, (10)(10), 32-34 (1983-10-01)
[Poisoning with liquid THF-M].
T N Guliaeva et al.
Sudebno-meditsinskaia ekspertiza, 24(2), 23-25 (1981-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service