Skip to Content
Merck
All Photos(1)

Documents

89025

Sigma-Aldrich

Copper(I) thiophenolate

≥98.0% (RT)

Synonym(s):

PhSCu, Copper(I) thiophenoxide, Phenylthiocopper(I), Thiophenol copper(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCu
CAS Number:
Molecular Weight:
172.71
Beilstein:
3709171
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (RT)

form

solid

reaction suitability

core: copper
reagent type: catalyst

mp

280 °C (dec.) (lit.)

SMILES string

[Cu]Sc1ccccc1

InChI

1S/C6H6S.Cu/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1

InChI key

PNNJYDODNCALKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reagents used as the source of nucleophilic phenyl sulfide for preparation of lithiumalkyl(phenylthio)cuprates and organophosphinecopper(I) phenylthiolatecomplexes

Other Notes

Reagent for the simple preparation of lithium (phenylthio)alkyl-cuprates from lithium alkyls, used for conjugate addition to α,β-enones

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.A. Wender et al.
Tetrahedron Letters, 1245-1245 (1977)
Majetich, G.; Song, J. et al.
The Journal of Organic Chemistry, 58, 1030-1030 (1993)
E.J. Corey et al.
Tetrahedron Letters, 4597-4597 (1978)
E. Piers et al.
The Journal of Organic Chemistry, 40, 2694-2694 (1975)
G.H. Posner et al.
Synthesis, 662-662 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service