Skip to Content
Merck
All Photos(1)

Key Documents

88732

Sigma-Aldrich

(S)-1-{(RP)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine

≥97%

Synonym(s):

(1S)-1-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-2-[(1S)-1-[bis(3,5-dimethylphenyl)phosphino]ethyl]ferrocene (acc to CAS), Josiphos SL-J008-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H36F12FeP2
CAS Number:
Molecular Weight:
910.53
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Quality Level

Assay

≥97%

form

powder

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(c3cc(cc(c3)C(F)(F)F)C(F)(F)F)c4cc(cc(c4)C(F)(F)F)C(F)(F)F)P(c5cc(C)cc(C)c5)c6cc(C)cc(C)c6

InChI

1S/C39H31F12P2.C5H5.Fe/c1-21-9-22(2)12-30(11-21)52(31-13-23(3)10-24(4)14-31)25(5)34-7-6-8-35(34)53(32-17-26(36(40,41)42)15-27(18-32)37(43,44)45)33-19-28(38(46,47)48)16-29(20-33)39(49,50)51;1-2-4-5-3-1;/h6-20,25H,1-5H3;1-5H;/t25-;;/m0../s1

InChI key

OPQIIHGMWPUTDC-WLOLSGMKSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service