Skip to Content
Merck
All Photos(1)

Key Documents

88718

Sigma-Aldrich

(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine

≥97%

Synonym(s):

(2S)-1-[(1S)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS), (S)-(R)-Josiphos, Josiphos SL-J001-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$671.00
1 G
R$4,047.00
50 G
Inquire
100 G
Inquire

R$671.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$671.00
1 G
R$4,047.00
50 G
Inquire
100 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C36H44FeP2 · C2H5OH
CAS Number:
Molecular Weight:
640.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

R$671.00


Check Cart for Availability

Request a Bulk Order

description

ethanol adduct

Assay

≥97%

form

powder

optical purity

ee: ≥99%

functional group

phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)P(C5CCCCC5)C6CCCCC6

InChI

1S/C31H39P2.C5H5.Fe/c1-25(32(26-15-6-2-7-16-26)27-17-8-3-9-18-27)30-23-14-24-31(30)33(28-19-10-4-11-20-28)29-21-12-5-13-22-29;1-2-4-5-3-1;/h4-5,10-14,19-27H,2-3,6-9,15-18H2,1H3;1-5H;/t25-;;/m0../s1

InChI key

HGTBZFMPHBAUCQ-WLOLSGMKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service