Skip to Content
Merck
All Photos(1)

Key Documents

87307

Sigma-Aldrich

Tetrahexylammonium iodide

≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]4N(I)
CAS Number:
Molecular Weight:
481.58
Beilstein:
4164997
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (AT)

form

powder

mp

99-101 °C

SMILES string

[I-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.HI/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;/h5-24H2,1-4H3;1H/q+1;/p-1

InChI key

VRKHAMWCGMJAMI-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M W J Bandara et al.
Physical chemistry chemical physics : PCCP, 22(22), 12532-12543 (2020-05-27)
A series of highly efficient quasi-solid-state dye-sensitized solar cells (DSCs) is prepared by harnessing the binary cation effect and positive effects of the selected performance enhancers of gel-polymer electrolytes. The new electrolyte is composed of polyacrylonitrile polymer, tetra-hexylammonium iodide (Hex4NI)
K M Sunasara et al.
Biotechnology progress, 17(5), 897-906 (2001-10-06)
This work investigates the utility of RPLC displacement chromatography for the purification of recombinant brain derived neurotrophic factor (rHu-BDNF) from its variants and E. coli. protein (ECP) impurities. The closely associated variants (six in total) differ by one amino acid
L Zhang et al.
The Journal of pharmacology and experimental therapeutics, 288(3), 1192-1198 (1999-02-23)
Polyspecific organic cation transporters in epithelia play an important role in the elimination of many endogenous bioactive amines and therapeutically important drugs. Recently, the first human organic cation transporter (hOCT1) was cloned from liver. The purpose of the current study
A F Hofmann
Journal of lipid research, 8(1), 55-58 (1967-01-01)
Ethyl acetate or chloroform solutions of tetraheptylammonium chloride, an oil-soluble quaternary amine, quantitatively extract polar, anionic lipids such as steroid or bile salt conjugates from aqueous solution by a process of anion exchange.
Y Uyama et al.
Pflugers Archiv : European journal of physiology, 426(5), 363-370 (1994-03-01)
Effects of tetraalkylammonium ions, having tetraalkyl chains of increasing length from ethyl to octyl, on inositol-trisphosphate (InsP3)-induced Ca2+ release and contractile mechanics were examined in guinea-pig skinned ileal smooth muscle longitudinal strips. Although tetrahexylammonium ions (THexA) appeared to be the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service