Skip to Content
Merck
All Photos(1)

Key Documents

86929

Sigma-Aldrich

Tetrabutylphosphonium methanesulfonate

≥98.0% (NT)

Synonym(s):

Methanesulfonic acid tetrabutylphosphonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4P(CH3SO3)
CAS Number:
Molecular Weight:
354.53
Beilstein:
6030085
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

crystals

mp

59-62 °C

SMILES string

CS([O-])(=O)=O.CCCC[P+](CCCC)(CCCC)CCCC

InChI

1S/C16H36P.CH4O3S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

DSQCNXSPLHDLED-UHFFFAOYSA-M

Other Notes

Ionic liquid at ambient temperature; more stable than the corresponding ammonium salt; Stationary phase for GC having unique selectivities

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.M. Pomaville et al
Journal of Chromatography A, 438, 1-1 (1988)
M. Badri et al.
Tetrahedron Letters, 33, 4435-4435 (1992)
Shuverthi De Sarkar et al.
The European physical journal. E, Soft matter, 43(8), 55-55 (2020-08-21)
The physical properties of an aqueous solution of a macromolecule primarily depend on its chemical structure and the mesoscopic aggregates formed by many of such molecules. Ionic liquids (ILs) are the macromolecules that have caught significant research interests for their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service