Skip to Content
Merck
All Photos(2)

Documents

86856

Sigma-Aldrich

Tetrabutylammonium hydrogen difluoride solution

technical, ~50% in methylene chloride (T)

Synonym(s):

TBABF, Tetrabutylammonium bifluoride, Tetrabutylammonium difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HF2)
CAS Number:
Molecular Weight:
281.47
Beilstein:
4729304
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical

form

liquid

concentration

~50% in methylene chloride (T)

impurities

~5% tetrabutylammonium chloride

refractive index

n20/D 1.442

storage temp.

2-8°C

SMILES string

[F-].F[H].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.2FH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;/h5-16H2,1-4H3;2*1H/q+1;;/p-1

InChI key

ZHBDKVWQJKYIFF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Used as an etchant for silica films

Reactant for:
  • Nucleophilic fluorination
  • Ring-opening fluorination reaction
  • Substitution reactions
Tetrabutylammonium hydrogen difluoride (TBABF) can be used as a reagent:
  • For the conversion of aromatic boronic acids and esters into corresponding tetrabutylammonium trifluoroborate salts. These salts are utilized as surrogates of boronic acids in Suzuki-Miyaura cross-coupling reactions.
  • In the nucleophilic fluorination of organic halides, tosylates, and triflates.
  • For the regioselective synthesis of fluorohydrins by ring-opening of terminal epoxides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrabutylammonium Bifluoride
Prakash GS and Pertusati F
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service