Skip to Content
Merck
All Photos(1)

Key Documents

861480

Sigma-Aldrich

Dihydrocytochalasin B γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H39NO5
CAS Number:
Molecular Weight:
481.62
MDL number:

SMILES string

CC(CCCC1CCC(=O)O1)C\C=C\C2C(O)C(=C)C(C)C3C(Cc4ccccc4)NC(=O)C23O

InChI

1S/C29H39NO5/c1-18(9-7-13-22-15-16-25(31)35-22)10-8-14-23-27(32)20(3)19(2)26-24(30-28(33)29(23,26)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14,18-19,22-24,26-27,32,34H,3,7,9-10,13,15-17H2,1-2H3,(H,30,33)/b14-8+

InChI key

VJQJWFGRYRQFRH-RIYZIHGNSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service