Skip to Content
Merck
All Photos(1)

Key Documents

86020

Sigma-Aldrich

Sulfacetamide sodium salt monohydrate

≥98.0% (dried material, T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N2NaO3S · H2O
CAS Number:
Molecular Weight:
254.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (dried material, T)

impurities

~7% water

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

Mode of action

enzyme | inhibits

SMILES string

NC1=CC=C(S(N([Na])C(C)=O)(=O)=O)C=C1.O

InChI

1S/C8H10N2O3S.Na.H2O/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;;/h2-5H,9H2,1H3,(H,10,11);;1H2/q;+1;/p-1

InChI key

IHCDKJZZFOUARO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margareta Ungureanu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 110(2), 456-459 (2007-09-07)
In the present study we emphasized the antituberculosis action of new sulphacetamide derivatives. In order o extend the research for obtaining antituberculosis substances, we decided to study the influence of the introducing of the thiourea and sulphamide groups in the
Zoe Diana Draelos
Journal of drugs in dermatology : JDD, 9(3), 234-236 (2010-03-18)
Prior to 1962, some of the most versatile drugs in dermatology were approved by the U.S. Food and Drug Administration (FDA) solely on the basis of safety. One of these is the combination 10% sodium sulfacetamide and 5% sulfur. Sodium
James Q Del Rosso
Cutis, 86(5 Suppl), 16-25 (2011-01-15)
Rosacea is a prevalent inflammatory skin disorder that affects approximately 16 million individuals in the United States. Although its exact etiology is unknown, basic science, histologic evidence, and clinical evidence suggest that it is inflammatory in nature. In this 12-week
M Gooderham
Skin therapy letter, 14(2), 1-3 (2009-03-31)
Many options exist for the treatment of rosacea, including topical and systemic therapies, laser and light-based therapies, and surgical procedures. A classification system for rosacea identifies 4 subtypes (i.e., erythematotelangiectatic, papulopustular, phymatous, and ocular), which may help guide therapeutic decision-making.
Ilknur Akyol-Salman et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 28(1), 49-52 (2011-07-15)
This study aimed to compare the efficacy of topical N-acetyl-cysteine (NAC) with a topical steroid-antibiotic combination, betamethasone-sulfacetamide sodium therapy in patients with meibomian gland dysfunction (MGD). Twenty patients with MGD were prospectively randomized and assigned into 2 groups. The patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service