Skip to Content
Merck
All Photos(1)

Key Documents

808490

Sigma-Aldrich

N6-Propargyl-ATP sodium salt

Synonym(s):

(N6pATP) sodium salt, N6-Propargyl-adenosine-5’-triphosphate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,177.00

R$1,177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$1,177.00

About This Item

Empirical Formula (Hill Notation):
C13H18N5O13P3 · xNa+
Molecular Weight:
545.23 (free acid basis)
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

R$1,177.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

reaction suitability

reaction type: click chemistry

solubility

10 mM Tris-HCl, pH 7.5: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@@H]([C@H]1O)[C@@H](COP(O)(OP(O)(OP(O)(O)=O)=O)=O)O[C@@H]1N2C=NC3=C2N=CN=C3NCC#C

InChI

1S/C13H18N5O13P3/c1-2-3-14-11-8-12(16-5-15-11)18(6-17-8)13-10(20)9(19)7(29-13)4-28-33(24,25)31-34(26,27)30-32(21,22)23/h1,5-7,9-10,13,19-20H,3-4H2,(H,24,25)(H,26,27)(H,14,15,16)(H2,21,22,23)/t7-,9-,10-,13?/m1/s1

InChI key

MUOIWXPACQCYQC-RJNFYWFKSA-N

Application

N6-Propargyl-ATP is suitable for in vitro AMPylation of proteins as well as in vitro polyadenylation of RNA. The resulting alkyne-functionalized protein[1][2] or RNA[3] can be processed using Cu(I)-catalyzed click chemistry to attach azide-labeled molecules. This gives options such as introduction of a biotin group for purification tasks, introduction of a fluorescent group for detection, or crosslinking to other azide-functionalized biomolecules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A new chemical handle for protein AMPylation at the host-pathogen interface.
Malgorzata Broncel et al.
Chembiochem : a European journal of chemical biology, 13(2), 183-185 (2012-01-04)
Markus Grammel et al.
Chembiochem : a European journal of chemical biology, 13(8), 1112-1115 (2012-04-20)
A versatile "clickable" nucleoside: Metabolic labeling of cells is useful in studying the dynamics of biological molecules. N(6) pA can be utilized by all three mammalian RNA polymerases, as well as poly(A) polymerase. Because of its alkyne modification, RNA labeled
Markus Grammel et al.
Journal of the American Chemical Society, 133(43), 17103-17105 (2011-09-29)
Protein AMPylation is an emerging post-translational modification, which plays key roles in bacterial pathogenesis and cell biology. Enzymes with AMPylation activity, referred to as AMPylators, have been identified in several bacterial pathogens and eukaryotes. To facilitate the study of this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service