Skip to Content
Merck
All Photos(2)

Key Documents

808458

Sigma-Aldrich

2-Hydroxy-5-N-methacrylamidobenzoic acid

Synonym(s):

2-hydroxy-5-[(2-methyl-1-oxo-2-propen-1-yl)amino] benzoic acid, N-Methacryloyl-5-aminosalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO4
CAS Number:
Molecular Weight:
221.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

>95% (HPLC)

form

powder

mp

206-211 °C

storage temp.

2-8°C

SMILES string

OC1=CC=C(NC(C(C)=C)=O)C=C1C(O)=O

InChI

1S/C11H11NO4/c1-6(2)10(14)12-7-3-4-9(13)8(5-7)11(15)16/h3-5,13H,1H2,2H3,(H,12,14)(H,15,16)

InChI key

XEYLAWVXYZUVDD-UHFFFAOYSA-N

Application

Used as an adhesive primer in dental resorative systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fujisawa et al.
Dental materials journal, 11(1), 17-25 (1992-06-01)
N-methacryloyloxy-5-aminosalicylic acid (MASA) has recently been used as an adhesive primer in restorative resin systems. To monitor the biological activity of MASA, we studied changes in NMR-chemical shifts (delta H) and the differential scanning calorimetry (DSC) phase transition temperature (Tm)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service