Skip to Content
Merck
All Photos(5)

Key Documents

806668

Sigma-Aldrich

Graphene nanoplatelets

powder, hydrophobic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder

composition

Carbon, >95 wt. %
Oxygen, <2 wt. %

bulk density

0.04 g/mL

Looking for similar products? Visit Product Comparison Guide

General description

Low dimensional carbon nanoplatelet with a large lateral dimension (>25μ) and few in plane defects. Highly hydrophobic. Few layer exfoliation in the medium can be obtained by standard methods. It has been used with success as electrical conductive additive in powder coating formulation among other properties.

Application

  • Graphene (nano)composite materials
  • Conductive coatings
  • Anti-corrosion coatings
  • Conductive Inks
  • Energy Storage
  • Electrode materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ashish Asthana et al.
ACS applied materials & interfaces, 6(11), 8859-8867 (2014-05-23)
Superhydrophobic surfaces resisting water penetration into their texture under dynamic impact conditions and offering simultaneously additional functionalities can find use in a multitude of applications. We present a facile, environmentally benign, and economical fabrication of highly electrically conductive, polymer-based superhydrophobic
Processing of nanographene platelets (NGPs) and NGP nanocomposites: a review
Jang BZ, and Zhamu A.
J. Mater. Sci., 43(15), 5092-5101 (2008)
Ladislav Kavan et al.
ACS nano, 5(1), 165-172 (2010-12-04)
Commercial graphene nanoplatelets in the form of optically transparent thin films on F-doped SnO(2) (FTO) exhibited high electrocatalytic activity toward I(3)(-)/I(-) redox couple, particularly in electrolyte based on ionic liquid (Z952). The charge-transfer resistance, R(CT), was smaller by a factor
Mohammad-Bagher Ebrahim-Habibi et al.
Scientific reports, 9(1), 1273-1273 (2019-02-06)
Investigation of non-covalent interaction of hydrophobic surfaces with the protein G (PrG) is necessary due to their frequent utilization in immunosensors and ELISA. It has been confirmed that surfaces, including carbonous-nanostructures (CNS) could orient proteins for a better activation. Herein

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service