803766
PtCo/graphene nanocomposite
10 mg/mL, dispersion in acetone
Synonym(s):
PtCo alloy decorated highly conductive graphene
About This Item
description
Loading Density: 10%-50% (PtCo nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)
form
dispersion
composition
acetone, ~80 wt. %
graphene, 10-15%
PtCo nanoparticle, 5-10%
concentration
10 mg/mL
thickness
1-10 nm , graphene
particle size
2-5 nm (PtCo nanocrystal)
storage temp.
2-8°C
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
41.0 °F
Flash Point(C)
5 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service