Skip to Content
Merck
All Photos(1)

Key Documents

803723

Sigma-Aldrich

Mn3O4/graphene nanocomposite

10 mg/mL, dispersion in acetone

Synonym(s):

Mn3O4 decorated highly conductive graphene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

description

Loading Density: 30%-70% (Mn3O4 nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)

form

dispersion

composition

acetone, ~80 wt. %
graphene, 3-8%
Mn3O4 nanoparticle, 4-9%

concentration

10 mg/mL

thickness

1-10 nm , graphene

particle size

5-25 nm (Mn3O4 nanocrystal)

storage temp.

2-8°C

Application

High capacity anode material for supercapacitors and lithium ion batteries.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mn3O4 nanoparticles embedded into graphene nanosheets: Preparation, characterization, and electrochemical properties for supercapacitors.
Wang et al.
Electrochimica Acta, 55(22), 6812-6817 (2010)
Hailiang Wang et al.
Journal of the American Chemical Society, 132(40), 13978-13980 (2010-09-22)
We developed two-step solution-phase reactions to form hybrid materials of Mn(3)O(4) nanoparticles on reduced graphene oxide (RGO) sheets for lithium ion battery applications. Selective growth of Mn(3)O(4) nanoparticles on RGO sheets, in contrast to free particle growth in solution, allowed

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service