Skip to Content
Merck
All Photos(1)

Key Documents

803715

Sigma-Aldrich

Fe3O4/graphene nanocomposite

10 mg/mL, dispersion in acetone

Synonym(s):

Fe3O4 decorated highly conductive graphene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

description

Loading Density: 30%-70% (Fe3O4 nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)

form

dispersion

composition

acetone, ~80 wt. %
Fe3O4 nanoparticle, 4-9%
graphene, 3-8%

concentration

10 mg/mL in acetone

thickness

1-10 nm , graphene

particle size

5-25 nm (Fe3O4 nanocrystal)

storage temp.

2-8°C

Application

  • Electromagnetic wave absorption materials.
  • Electro-magnetic device.
  • Anode material for lithium-ion batteries.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step solvothermal preparation of Fe3O4/graphene composites at elevated temperature and their application as anode materials for lithium-ion batteries.
Laiying J, et al.
Royal Society of Chemistry Advances, 4(104), 59981-59989 (2014)
Fe3O4-Graphene Nanocomposites with Improved Lithium Storage and Magnetism Properties.
Jing S, et al.
The Journal of Physical Chemistry C, 115(30), 14469-14477 (2011)

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service