Skip to Content
Merck
All Photos(1)

Key Documents

799556

Sigma-Aldrich

Polycaprolactone trimethacrylate

average Mn 950

Synonym(s):

PCLTMA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

Quality Level

mol wt

average Mn 950

refractive index

n20/D 1.476

density

1.120 at 25 °C

storage temp.

−20°C

Application

Polycaprolactone is a biodegradable polymer used in biomedical applications. Polycaprolactone trimethacrylate has been used as a building block for a biodegradable crosslinked polymer networks.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses and evaluation of biodegradable multifunctional polymer networks
Chung I, et al.
Eur. Polymer J., 39(9), 1817-1822 (2003)
A Novel Injectable Poly(e-caprolactone)/Calcium Sulfate System for Bone Regeneration: Synthesis and Characterization.
La Gatta A
Macromolecular Bioscience, 2, 1108-1117 (2005)

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service