Skip to Content
Merck
All Photos(1)

Key Documents

798061

Sigma-Aldrich

(3E)-4-Phenylbut-3-enoic acid methyl ester

Synonym(s):

Methyl (3E)-4-Phenylbut-3-enoate, Methyl (E)-4-phenylbut-3-enoate, Methyl trans-styryl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$364.00

R$364.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$364.00

About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$364.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

functional group

ester
phenyl

storage temp.

2-8°C

SMILES string

O=C(OC)C/C=C/C1=CC=CC=C1

InChI

1S/C11H12O2/c1-13-11(12)9-5-8-10-6-3-2-4-7-10/h2-8H,9H2,1H3/b8-5+

InChI key

VOEDRUGFNJEWGQ-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Building block is reported by Huw Davies and coworkers as a precursor to the 2013 EROS Best Reagent; Methyl (E)-4-Phenyl-2-diazo-3-butenoate. Methyl (E)-4-Phenyl-2-diazo-3-butenoate is a donor-acceptor precursor to the corresponding Ru-carbenoid that is utilized in C-H insertion and cyclopropanation reactions.

Preparation Note

Preparatory information to make (E)-4-Phenyl-2-diazo-3-butenoate can be found by clicking the link below under other notes.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service