795143
2,6-Dicyclopentylaniline
95%
Synonym(s):
2,6-Dicyclopentylaniline
About This Item
Assay
95%
refractive index
n/D 1.571
density
1.034 at 25 °C
SMILES string
NC1=C(C2CCCC2)C=CC=C1C3CCCC3
InChI
1S/C16H23N/c17-16-14(12-6-1-2-7-12)10-5-11-15(16)13-8-3-4-9-13/h5,10-13H,1-4,6-9,17H2
InChI key
IEIPZQFILTYQLK-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service