Skip to Content
Merck
All Photos(1)

Key Documents

78927

Sigma-Aldrich

(S)-(+)-1-Phenyl-2-propanol

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(+)-α-Methylphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O
CAS Number:
Molecular Weight:
136.19
Beilstein:
2553541
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D +41±1°, c = 5.3% in benzene

optical purity

enantiomeric ratio: ≥99:1 (GC)

refractive index

n20/D 1.521

bp

95-97 °C/11 mmHg (lit.)

density

0.993 g/mL at 20 °C (lit.)

SMILES string

C[C@H](O)Cc1ccccc1

InChI

1S/C9H12O/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3/t8-/m0/s1

InChI key

WYTRYIUQUDTGSX-QMMMGPOBSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service