Skip to Content
Merck
All Photos(1)

Key Documents

78240

Sigma-Aldrich

(S)-3-Phenylbutyric acid

≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

(S)-β-Methylhydrocinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Beilstein:
4663842
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
form:
liquid
Assay:
≥99.0% (sum of enantiomers, HPLC)

Assay

≥99.0% (sum of enantiomers, HPLC)

form

liquid

optical activity

[α]20/D +57±2°, c = 1% in benzene

refractive index

n20/D 1.518

bp

94-95 °C/0.3 mmHg (lit.)

density

1.069 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](CC(O)=O)c1ccccc1

InChI

1S/C10H12O2/c1-8(7-10(11)12)9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,11,12)/t8-/m0/s1

InChI key

ZZEWMYILWXCRHZ-QMMMGPOBSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service