Skip to Content
Merck
All Photos(1)

Documents

778443

Sigma-Aldrich

2,4,6-Triisopropylbenzenesulfonyl azide solution

10%±2% in toluene, ≥98% (HPLC)

Synonym(s):

Trisyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O2S
CAS Number:
Molecular Weight:
309.43
Beilstein:
2391566
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (HPLC)

form

liquid

concentration

10%±2% in toluene

impurities

≤0.015% water

refractive index

n20/D 1.499

storage temp.

2-8°C

SMILES string

CC(C)c1cc(C(C)C)c(c(c1)C(C)C)S(=O)(=O)N=[N+]=[N-]

InChI

1S/C15H23N3O2S/c1-9(2)12-7-13(10(3)4)15(14(8-12)11(5)6)21(19,20)18-17-16/h7-11H,1-6H3

InChI key

AEMWUHCKKDPRSK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service