Skip to Content
Merck
All Photos(1)

Key Documents

777927

Sigma-Aldrich

2-(4-Morpholino)ethyl 2-bromoisobutyrate

contains MEHQ as inhibitor, 95% (excluding up to 10% residual Toluene)

Synonym(s):

2-Bromo-2-methylpropanoic acid 2-(4-morpholinyl)ethyl ester, 2-Morpholinoethyl 2-bromo-2-methylpropanoate, ATRP initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18BrNO3
CAS Number:
Molecular Weight:
280.16
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95% (excluding up to 10% residual Toluene)

form

liquid

contains

MEHQ as inhibitor

refractive index

n/D 1.486

density

1.258 at 25 °C

storage temp.

2-8°C

SMILES string

BrC(C)(C)C(OCCN1CCOCC1)=O

InChI

1S/C10H18BrNO3/c1-10(2,11)9(13)15-8-5-12-3-6-14-7-4-12/h3-8H2,1-2H3

InChI key

XEXMMWOKORAPHR-UHFFFAOYSA-N

Application

This bromoisobutyrate is commonly used as an ATRP initiator. The morpholino functionality can be used to bind to RNA or other therapeutics.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Synthesis of Well-Defined Quaternized Homopolymers and Diblock Copolymers via ATRP in Protic Media.
Li Y, et al.
Macromolecules, 36, 8268-8268 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service