776637
Benzylmagnesium chloride solution
1.0 M in 2-methyltetrahydrofuran
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H5CH2MgCl
CAS Number:
Molecular Weight:
150.89
Beilstein:
878506
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solution
reaction suitability
reaction type: Grignard Reaction
concentration
1.0 M in 2-methyltetrahydrofuran
density
0.899 g/mL at 25 °C
SMILES string
Cl[Mg]Cc1ccccc1
InChI
1S/C7H7.ClH.Mg/c1-7-5-3-2-4-6-7;;/h2-6H,1H2;1H;/q;;+1/p-1
InChI key
NRAFPLGJPPJUNB-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF)
Packaging
The 25 mL Sure/Seal™ bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.
Legal Information
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
8.6 °F - closed cup
Flash Point(C)
-13.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service