Skip to Content
Merck
All Photos(2)

Documents

776092

Sigma-Aldrich

N,N-Dibenzyl-(2-pyridinecarboxaldehyde)hydrazone

97%

Synonym(s):

2-[(2,2-Dibenzylhydrazono)methyl]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N3
CAS Number:
Molecular Weight:
301.38
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

65-71 °C

SMILES string

C(N(Cc1ccccc1)\N=C\c2ccccn2)c3ccccc3

InChI

1S/C20H19N3/c1-3-9-18(10-4-1)16-23(17-19-11-5-2-6-12-19)22-15-20-13-7-8-14-21-20/h1-15H,16-17H2/b22-15+

InChI key

IPXKRUCFNUYSKI-PXLXIMEGSA-N

Related Categories

Application

Nitrogen-Directed Iridium-Catalyzed Borylations of Arenes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of hemilabile N,N ligands in nitrogen-directed iridium-catalyzed borylations of arenes.
Abel Ros et al.
Angewandte Chemie (International ed. in English), 50(49), 11724-11728 (2011-10-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service