Skip to Content
Merck
All Photos(1)

Key Documents

773077

Sigma-Aldrich

Fmoc-N-Me-Asp(OtBu)-OH

97%, for peptide synthesis

Synonym(s):

N-Methyl aspartic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$3,048.00

R$3,048.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$3,048.00

About This Item

Empirical Formula (Hill Notation):
C24H27NO6
CAS Number:
Molecular Weight:
425.47
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

R$3,048.00


Check Cart for Availability

Request a Bulk Order

Product Name

Fmoc-N-Me-Asp(OtBu)-OH, 97%

Assay

97%

form

powder or crystals

optical activity

[α]22/D -39.0°, c = 0.5% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

135-140 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC([C@@H](N(C(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O)C)CC(OC(C)(C)C)=O)=O

InChI

1S/C24H27NO6/c1-24(2,3)31-21(26)13-20(22(27)28)25(4)23(29)30-14-19-17-11-7-5-9-15(17)16-10-6-8-12-18(16)19/h5-12,19-20H,13-14H2,1-4H3,(H,27,28)/t20-/m0/s1

InChI key

CYWWLVIEAOUXGW-FQEVSTJZSA-N

Application

Fmoc protected N-methyl amino acid suitable for solid phase peptide synthesis. N-methyl amino acids have been shown to improve proteolytic stability of peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service