Skip to Content
Merck
All Photos(1)

Key Documents

771538

Sigma-Aldrich

Yttrium 2-methoxyethoxide solution

15-20 wt. % in 2-methoxyethanol

Synonym(s):

Y(OEtOMe)3, Yttrium trimethoxyethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21O6Y
Molecular Weight:
314.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: yttrium

concentration

15-20 wt. % in 2-methoxyethanol
~5 wt. % Y

refractive index

n20/D 1.416

density

1.039 g/mL at 25 °C

SMILES string

COCCO[Y](OCCOC)OCCOC

InChI

1S/3C3H7O2.Y/c3*1-5-3-2-4;/h3*2-3H2,1H3;/q3*-1;+3

InChI key

IVXUPDRNSUCVNX-UHFFFAOYSA-N

Application

Yttrium methoxyethoxide; Y(OEtOMe)3 is an important precursor for the sol-gel preparation of Y2O3-containing materials; example phosphor materials like YAG; yttria stabilized zirconia (YSZ) powders and films for solid oxide fuel cells.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chong M. K.;
Materials Chemistry and Physics, 100(2-3), 329-329 (2006)
Suarez; M.;
J. Alloy Compounds, 493(1-2), 391-391 (2010)

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service