768669
4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine
97%
Synonym(s):
2-TNATA, 2TNATA
About This Item
Recommended Products
Assay
97%
form
powder
mp
241-248 °C
λmax
329 nm in chloroform
SMILES string
c1ccc(cc1)N(c2ccc(cc2)N(c3ccc(cc3)N(c4ccccc4)c5ccc6ccccc6c5)c7ccc(cc7)N(c8ccccc8)c9ccc%10ccccc%10c9)c%11ccc%12ccccc%12c%11
InChI
1S/C66H48N4/c1-4-22-55(23-5-1)68(64-31-28-49-16-10-13-19-52(49)46-64)61-40-34-58(35-41-61)67(59-36-42-62(43-37-59)69(56-24-6-2-7-25-56)65-32-29-50-17-11-14-20-53(50)47-65)60-38-44-63(45-39-60)70(57-26-8-3-9-27-57)66-33-30-51-18-12-15-21-54(51)48-66/h1-48H
InChI key
KDOQMLIRFUVJNT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service