Skip to Content
Merck
All Photos(2)

Key Documents

768634

Sigma-Aldrich

Zirconium(IV) 2-ethylhexanoate in mineral spirits

~6% Zr basis

Synonym(s):

Zirconium octoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(C8H15O2)4
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: zirconium

concentration

~6% Zr

refractive index

n20/D 1.446

bp

167-178 °C/760 mmHg

density

0.870 g/mL at 25 °C

SMILES string

CCCCC(CC)C(=O)O[Zr](OC(=O)C(CC)CCCC)(OC(=O)C(CC)CCCC)OC(=O)C(CC)CCCC

InChI

1S/4C8H16O2.Zr/c4*1-3-5-6-7(4-2)8(9)10;/h4*7H,3-6H2,1-2H3,(H,9,10);/q;;;;+4/p-4

InChI key

OFYFURKXMHQOGG-UHFFFAOYSA-J

Application

Zirconium 2-ethylhexanoate is a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium; example: zirconium oxide thin films; yttria stabilized zirconia films for solid oxide fuel cells.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Miyamoto;
Applied Surface Science, 197-198, 398-398 (2002)
Stollberg; D. W.;
Thin Solid Films, 483(1-2), 211-211 (2005)

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service