Skip to Content
Merck
All Photos(2)

Key Documents

766755

Sigma-Aldrich

2,6-Diaminopurine-9-arabinoside

97%

Synonym(s):

2,6-Diamino-9-(β-D-arabinofuranosyl)purine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N6O4
CAS Number:
Molecular Weight:
282.26
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D +28.5°, c = 0.5% in hydrochloric acid

mp

286-290 °C

storage temp.

2-8°C

SMILES string

Nc1nc(N)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)c2n1

InChI

1S/C10H14N6O4/c11-7-4-8(15-10(12)14-7)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H4,11,12,14,15)/t3-,5-,6+,9-/m1/s1

InChI key

ZDTFMPXQUSBYRL-FJFJXFQQSA-N

Application

Versatile nucleoside intermediate.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Antiviral effects of 9-beta-D-arabinofuranosyl-2,6-diaminopurine against experimental herpetic keratitis].
S Yamane
Nippon Ganka Gakkai zasshi, 88(3), 586-594 (1984-03-01)
[Comparative studies of anti-herpes simplex virus compounds in vitro].
S Inoue et al.
Nippon Ganka Gakkai zasshi, 86(11), 2113-2121 (1982-01-01)
S Bajaj et al.
Blood, 62(1), 75-80 (1983-07-01)
We compared the effect of adenosine and adenosine analogues on the phytohemagglutinin-induced proliferative response of blood lymphocytes from normal subjects and patients with chronic lymphocytic leukemia. As measured by the inhibition of thymidine or leucine incorporation, adenosine was more toxic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service