Skip to Content
Merck
All Photos(1)

Key Documents

765147

Sigma-Aldrich

2-Nitro-5-(2-propynyloxy)benzyl 4-cyano-4-(phenylcarbonothioylthio)pentanoate

97%

Synonym(s):

Clickable CTA, Light-sensitive clickable RAFT agent, RAFT agent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N2O5S2
CAS Number:
Molecular Weight:
468.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

90-94 °C

storage temp.

2-8°C

SMILES string

C#CCOC1=CC(COC(CCC(C)(C#N)SC(C2=CC=CC=C2)=S)=O)=C([N+]([O-])=O)C=C1

InChI

1S/C23H20N2O5S2/c1-3-13-29-19-9-10-20(25(27)28)18(14-19)15-30-21(26)11-12-23(2,16-24)32-22(31)17-7-5-4-6-8-17/h1,4-10,14H,11-13,15H2,2H3

InChI key

IOYDTDXAMFFKOA-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of methacrylate and methacrylamide monomers. Chain Transfer Agent (CTA) with alkyne functionality for ligation and a photo-sensitive moiety for controlled cleavage.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lida Ahmadkhani et al.
Artificial cells, nanomedicine, and biotechnology, 46(5), 1052-1063 (2017-07-30)
A drug delivery system based on dual responsive units was developed. An appealing pH- and thermo-responsive triblock terpolymer as the drug carrier was synthesized by RAFT polymerization of N-isopropyl acrylamide and methacrylic acid monomers using PEG-RAFT agent. The Fe3O4 magnetic
Mirim Park et al.
Macromolecular rapid communications, 41(20), e2000399-e2000399 (2020-09-10)
Redox-initiated reversible addition-fragmentation chain transfer (RAFT) miniemulsion polymerizations are successfully conducted with an employment of trithiocarbonate-based macro-RAFT agents and surfactant. Two macro-RAFT agents-hydrophilic poly(poly(ethylene glycol) methyl ether methacrylate) (PPEGMA27 ) and amphiphilic poly(poly(ethylene glycol) methyl ether methacrylate)-b-polystyrene (PPEGMA27 -b-PS33 )-
Thomas Swift et al.
Soft matter, 12(9), 2542-2549 (2016-01-30)
Fluorescence spectroscopy on a series of aqueous solutions of poly(acrylic acid) containing a luminescent label showed that polymers with molar mass, Mn < 16.5 kDa did not exhibit a pH responsive conformational change, which is typical of higher molar mass
Shunsuke Tazawa et al.
Macromolecular rapid communications, 41(19), e2000346-e2000346 (2020-08-19)
Thermoplastic solid poly(2-methoxyethyl acrylate) (PMEA)-based polyurethane (PU) is synthesized through the reversible addition-fragmentation chain transfer (RAFT) polymerization and the condensation polymerization, using hydroxyl-terminated RAFT reagents and diisocyanate, respectively. Neat PMEA is a promising antithrombogenic liquid used in the medical fields.
Zahra Abousalman-Rezvani et al.
Carbohydrate polymers, 225, 115247-115247 (2019-09-16)
Cellulose nanocrystals (CNC)-grafted and free copolymers were synthesized in three different ratios of DMAEMA and coumarin monomers (30:5, 40:7, and 50:10) through reversible addition-fragmentation chain transfer polymerization. These multi-responsive polymers to carbon dioxide (CO2), temperature, and light triggers can be

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service