Skip to Content
Merck
All Photos(1)

Key Documents

765015

Sigma-Aldrich

2,2′-Dimethyl-N,N′-di-[(1-naphthyl)-N,N′-diphenyl]-1,1′-biphenyl-4,4′-diamine

97% (HPLC)

Synonym(s):

α-NPD, N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)-2,2′-dimethylbenzidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H36N2
CAS Number:
Molecular Weight:
616.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (HPLC)

form

solid

mp

247-252 °C

λmax

306 nm in THF

SMILES string

Cc1cc(ccc1-c2ccc(cc2C)N(c3ccccc3)c4cccc5ccccc45)N(c6ccccc6)c7cccc8ccccc78

InChI

1S/C46H36N2/c1-33-31-39(47(37-19-5-3-6-20-37)45-25-13-17-35-15-9-11-23-43(35)45)27-29-41(33)42-30-28-40(32-34(42)2)48(38-21-7-4-8-22-38)46-26-14-18-36-16-10-12-24-44(36)46/h3-32H,1-2H3

InChI key

ZJFKMIYGRJGWIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2′-Dimethyl-N,N′-di-[(1-naphthyl)-N,N′-diphenyl]-1,1′-biphenyl-4,4′-diamine (α-NPD) is a conducting polymer that can be used as a hole transporting material (HTM) with a transport gap of ~4 eV and an ionization potential of 5.5 eV.[1]

Application

α-NPD can be used as a π-conjugated organic semiconducting material which has good charge transporting properties. It can be used in the fabrication of organic electronic devices such organic light emitting diodes (OLEDs) and organic solar cells (OSCs).[2][3][4]
This material is used as a high performance host material for OLED devices showing increased device efficiency (maximum luminance efficiency was 4.4 cd/A at 9 V for the fluorescent device and 24.4 cd/A for phosphorescent device).[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical band gaps of organic semiconductor materials
Costa JCS, et al.
Optical Materials, 58(1), 51-60 (2016)
Determination of charge-carrier transport in organic devices by admittance spectroscopy: Application to hole mobility in alpha-NPD
Nguyen ND, et al.
Physical Review. B, Condensed Matter and Materials Physics, 75(7), 075307-075307 (2007)
Tsai; Y.- S.; et al.
Thin Solid Films, 517, 5338-5338 (2009)
Band alignment at organic-inorganic semiconductor interfaces: alpha-NPD and CuPc on InP (110)
Chasse T, et al.
Journal of Applied Physics, 85(9), 6589-6592 (1999)
Impact of electrode contamination on the alpha-NPD/Au hole injection barrier
Wan A, et al.
Organic Electronics, 6(1), 47-54 (2005)

Articles

Highly reducing or oxidizing species enhance organic semiconductor conductivity by reducing charge-carrier injection barriers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service