Skip to Content
Merck
All Photos(1)

Documents

764736

Sigma-Aldrich

Poly(ethylene glycol) methyl ether-block-poly(D,L lactide)-block-decane

PEG average Mn 2,000, PDLLA average Mn 2,000

Synonym(s):

PEG-PDLLA-decane, PEG-b-PLA-b-decane, PEG-PLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

mol wt

PDLLA average Mn 2,000
PEG average Mn 2,000
average Mn 4,000 (total)

degradation timeframe

2-5 weeks

transition temp

Tm 29-33 °C

PDI

<1.1 (typical PEG)
<1.2
<1.3 (overall)

storage temp.

2-8°C

General description

Block copolymer micelles are widely used in drug delivery applications. PEG-PDLLA is a biodegradable polymeric micelle which is used as a carrier for hydrophobic drugs like Paclitaxel. The hydrophilic PEG and hydrophobic PDLLA form the micelle core wherein the hydrophobic drug is loaded. The incorporation of the 10-alkyl end cap (decane) increases the hydrophobicity of the micelle core and increases its solubilizing capability for hydrophobic drugs.†

Application

Used as a carrier for the controlled and targeted release of anticancer hydrophobic drugs.

Features and Benefits

  • Good biocompatibility, low immunogenicity and good degradability.
  • Properties can be easily modulated by changing the block copolymer segment sizes to suit a particular application.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arunvel Kailasan et al.
Acta biomaterialia, 6(3), 1131-1139 (2009-09-01)
This work describes the synthesis and characterization of novel thermoresponsive highly branched polyamidoamine-polyethylene glycol-poly(D,L-lactide) (PAMAM-PEG-PDLLA) core-shell nanoparticles. A series of dendritic PEG-PDLLA nanoparticles were synthesized through conjugation of PEG of various chain lengths (1500, 6000 and 12,000 g mol(-1)) to
Hongtao Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(18), 6596-6601 (2008-05-01)
It is generally assumed that polymeric micelles, upon administration into the blood stream, carry drug molecules until they are taken up into cells followed by intracellular release. The current work revisits this conventional wisdom. The study using dual-labeled micelles containing

Articles

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service