Skip to Content
Merck
All Photos(1)

Key Documents

761745

Sigma-Aldrich

5-bromobenzothiazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$600.00

R$600.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$600.00

About This Item

Empirical Formula (Hill Notation):
C7H4BrNS
CAS Number:
Molecular Weight:
214.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$600.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

105-110 °C

functional group

bromo

SMILES string

Brc1ccc2scnc2c1

InChI

1S/C7H4BrNS/c8-5-1-2-7-6(3-5)9-4-10-7/h1-4H

InChI key

KFDDRUWQFQJGNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service