Skip to Content
Merck
All Photos(1)

Key Documents

760226

Sigma-Aldrich

2-Pyridinepropionic acid

97%

Synonym(s):

3-(2-Pyridyl)propionic acid, 3-(Pyridin-2-yl)propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,212.00

R$1,212.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Select a Size

Change View
1 G
R$1,212.00

About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,212.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

140-145 °C

functional group

carboxylic acid

SMILES string

n1c(cccc1)CCC(=O)O

InChI

1S/C8H9NO2/c10-8(11)5-4-7-3-1-2-6-9-7/h1-3,6H,4-5H2,(H,10,11)

InChI key

AZYKGQOIAGPVCK-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service