Skip to Content
Merck
All Photos(1)

Key Documents

76016

Sigma-Aldrich

Dichloro-methyl-octadecylsilane

Wacker Chemie AG, ≥92% n- and iso-isomer basis (GC)

Synonym(s):

Silane ODM, Methyl-octadecyl-dichlorosilane, Octadecyl-methyl-dichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H40Cl2Si
CAS Number:
Molecular Weight:
367.51
Beilstein:
1774816
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥85% n-isomer basis
≥92% n- and iso-isomer basis (GC)

form

liquid

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.457 (lit.)

bp

185 °C/2.5 mmHg (lit.)

mp

~25 °C (lit.)

density

0.92 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](C)(Cl)Cl

InChI

1S/C19H40Cl2Si/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(2,20)21/h3-19H2,1-2H3

InChI key

GYWBHBXGYTYXRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dichloro-methyl-octadecylsilane can be used to:
  • Prepare dipyridylsilane receptors by reacting with pyridinemethanol using triethylamine at RT.
  • Functionalize a new mesoporous silica material used for Li-ion batteries.
  • Prepare papers having hydrophobic self-assembled monolayers.

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>302.0 °F - DIN 51758

Flash Point(C)

> 150 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of hydrophobic self-assembled monolayers on paper surface with silanes
Oh M, et al.
Journal of Industrial and Engineering Chemistry, 17(1), 149-153 (2011)
Pyridylmethylsilanes as dicarboxyacid receptors: Experimental and theoretical study
Leska B, et al.
Journal of Molecular Structure, 1027, 116-123 (2012)
Composite polymer electrolytes with modified mesoporous silica filler for Li ion batteries
Osinska-Broniarz M, et al.
Composites Theory and Practice, 15(3), 124-129 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service