Skip to Content
Merck
All Photos(3)

Key Documents

75993

Sigma-Aldrich

Palladium on activated charcoal

greener alternative

moistened with water, 10% Pd basis (based on dry substance)

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$1,417.00
50 G
R$4,578.00

R$1,417.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$1,417.00
50 G
R$4,578.00

About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22

R$1,417.00


Check Cart for Availability

Request a Bulk Order

form

paste

quality

moistened with water

contains

~50% water as stabilizer

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Designing Safer Chemicals
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10% Pd

greener alternative category

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for hydrogenation of alkenes, alkynes, ketones, nitriles, imines, azides, nitro groups, benzenoid and heterocyclic aromatics; used for hydrogenolysis of cyclopropanes, benzyl derivatives, epoxides, hydrazines, and halides; used to dehydrogenate aromatics and deformylate aldehydes.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of GSKDevelopment of GSK's reagent guides ? embedding sustainability into reagent selections reagent guides ? embedding sustainability into reagent selection.
Adams JP, et al.
Green Chemistry, 15, 1542-1549 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service