Skip to Content
Merck
All Photos(1)

Key Documents

758345

Sigma-Aldrich

2-Chloro-3-(hydroxymethylene)-1-cyclohexene-1-carboxaldehyde

≥95%

Synonym(s):

2-Chloro-1-formyl-3-(hydroxymethylene)-1-cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9ClO2
CAS Number:
Molecular Weight:
172.61
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95%

form

solid

mp

113-116 °C (lit.)

storage temp.

−20°C

SMILES string

ClC1=C(C([H])=O)CCC/C1=C([H])/O

InChI

1S/C8H9ClO2/c9-8-6(4-10)2-1-3-7(8)5-11/h4-5,10H,1-3H2/b6-4-

InChI key

JWUQFQYYMGMPKE-XQRVVYSFSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jing Xing et al.
Journal of nanoscience and nanotechnology, 20(2), 668-672 (2019-08-07)
Currently, optical probes with near-infrared (NIR) fluorescence are of great interest in chemical biology. In the present study, we designed and synthesized a novel NIR fluorescent probe, IR789. IR789 has high selectivity and sensitivity for living cells imaging. The stronger

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service