Skip to Content
Merck
All Photos(1)

Documents

758167

Sigma-Aldrich

2-ethynylthiophene

90%

Synonym(s):

2-Acetylenylthiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4S
CAS Number:
Molecular Weight:
108.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.580

density

1.139 g/mL at 25 °C

storage temp.

−20°C

SMILES string

C#Cc1cccs1

InChI

1S/C6H4S/c1-2-6-4-3-5-7-6/h1,3-5H

InChI key

LWISLHRIEATKTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

108.0 °F

Flash Point(C)

42.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service