Skip to Content
Merck
All Photos(3)

Key Documents

757152

Sigma-Aldrich

5-Bromoisophthalic acid

greener alternative

97%

Synonym(s):

5-Bromo-1,3-benzenedicarbonic acid, 5-Bromo-1,3-benzenedicarboxylic acid, 5-Bromoisophthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5BrO4
CAS Number:
Molecular Weight:
245.03
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

270-275 °C
281-286 °C

greener alternative category

SMILES string

OC(=O)c1cc(Br)cc(c1)C(O)=O

InChI

1S/C8H5BrO4/c9-6-2-4(7(10)11)1-5(3-6)8(12)13/h1-3H,(H,10,11)(H,12,13)

InChI key

JATKASGNRMGFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Functionalizable MOF linker with an electron rich hetero atom.
Reactant for:
  • Preparation of manganese(II) bromoisophthalate bis(pyridyl)ethane coordination polymer
  • Preparation of nickel(II) and cobalt(II) bromoisophthalate coordination polymers
  • Preparation of copper bromoisophthalate bis(triazolyl)butane coordination polymer
  • Preparation of cesium isophthalate crown ether complexes
  • Hydrothermal preparation of zinc and cadmium bromoisophthalate polymeric complexes with dipyridyl-type coligands

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyasu Furukawa et al.
Journal of the American Chemical Society, 130(35), 11650-11661 (2008-08-13)
Metal-organic polyhedra and frameworks (MOPs and MOFs) were prepared by linking square units M2(CO2)4 (M = Cu and Zn) with a variety of organic linkers designed to control the dimensionality (periodicity) and topology of the resulting structures. We describe the
Syntheses and characterization of nickel(II) and cobalt(II) coordination polymers based on 5-bromoisophthalate anion and bis(imidazole) ligands
Ma, L.-F.; et al.
CrystEngComm, 13, 4625-4634 (2011)
Liu; G-X.; et al.
Journal of Chemical Crystallography, 40, 283-283 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service