Skip to Content
Merck
All Photos(2)

Key Documents

755141

Sigma-Aldrich

Carbon nanotube, double-walled

short, ≤10% Metal Oxide(TGA)

Synonym(s):

NC-2150, DWNT, Double wall carbon nanotubes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder

avg. diam. × L

3.5 nm × 3 μm , TEM

impurities

≤10% Metal Oxide (TGA)

Looking for similar products? Visit Product Comparison Guide

General description

Structure of double walled carbon nanotubes (DWNTs) can be imagined as a two coaxial concentric single walled carbon nanotubes (SWNTs). It has been prepared by catalytic chemical vapor deposition. DWNTs exhibit excellent field emission properties and may be potentially useful as field emission displays (FEDs) and field emission transistors (FETs).

Application

Applications include:
  • Electronics
  • Sensors
  • Composites
  • Energy Storage
  • Study of Life Science systems

Preparation Note

Catalytic Carbon Vapor Deposition (CCVD) Method

Legal Information

Product of Nanocyl, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gram-scale CCVD synthesis of double-walled carbon nanotubes.
Flahaut E, et al.
Chemical Communications (Cambridge, England), 12, 1442-1443 (2003)
Size-selective growth of double-walled carbon nanotube forests from engineered iron catalysts.
Yamada T, et al.
Nature Nanotechnology, 1(2), 131-136 (2006)
Wai Li Ling et al.
ACS nano, 5(2), 730-737 (2011-01-11)
The classical pathway of complement is an essential component of the human innate immune system involved in the defense against pathogens as well as in the clearance of altered self-components. Activation of this pathway is triggered by C1, a multimolecular
Amin Salehi-Khojin et al.
ACS nano, 5(1), 153-158 (2010-12-29)
There has been recent controversy whether the response seen in carbon nanotube (CNT) chemiresistors is associated with a change in the resistance of the individual nanotubes or changes in the resistance of the junctions. In this study, we carry out
Dominick J Bindl et al.
ACS nano, 4(10), 5657-5664 (2010-10-07)
Semiconducting single-walled carbon nanotubes (s-SWCNTs) have strong near-infrared and visible absorptivity and exceptional charge transport characteristics, rendering them highly attractive semiconductor absorbers for photovoltaic and photodetector technologies. However, these applications are limited by a poor understanding of how photogenerated charges

Articles

Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service