Skip to Content
Merck
All Photos(1)

Key Documents

754889

Sigma-Aldrich

Spiro-2CBP

≥98%

Synonym(s):

Spiro-2CBP, 2,7-Bis(N-carbazolyl)-9,9-spirofluorene, 2,7-Bis(carbazol-9-yl)-9,9-spirobifluorene, 9,9′-(9,9′-Spirobi[9H-fluorene]-2,7-diyl)bis-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H30N2
CAS Number:
Molecular Weight:
646.78
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

form

powder or crystals

mp

391-396 °C

absorption

110,000 at 231 nm in THF
33,400 at 293 nm in THF
41,500 at 343 nm in THF

fluorescence

λex 292 nm; λem 366 nm in THF

Orbital energy

HOMO 5.03 eV 
LUMO 1.65 eV 

SMILES string

c1ccc2c(c1)-c3ccccc3C24c5cc(ccc5-c6ccc(cc46)-n7c8ccccc8c9ccccc79)-n%10c%11ccccc%11c%12ccccc%10%12

InChI

1S/C49H30N2/c1-7-19-41-33(13-1)34-14-2-8-20-42(34)49(41)43-29-31(50-45-21-9-3-15-37(45)38-16-4-10-22-46(38)50)25-27-35(43)36-28-26-32(30-44(36)49)51-47-23-11-5-17-39(47)40-18-6-12-24-48(40)51/h1-30H

InChI key

UDECBOWBCXTHEY-UHFFFAOYSA-N

Application

This material is a popular phosphorescent host material for OLED devices.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of a polarity matching and high-energy exciton generating host in fabricating efficient purplish-blue OLEDs from a sky-blue emitter
Jou, J-H; et al.
Journal of Materials Chemistry, 22, 15500-15506 (2012)
Francesco Rodella et al.
Frontiers in chemistry, 8, 657-657 (2020-08-28)
The synthesis of stable blue TADF emitters and the corresponding matrix materials is one of the biggest challenges in the development of novel OLED materials. We present six bipolar host materials based on triazine as an acceptor and two types

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service