Skip to Content
Merck
All Photos(1)

Documents

752371

Sigma-Aldrich

Poly(ethylene glycol), 16 hydroxyl dendron, generation 3

average Mn 8,000

Synonym(s):

PFD-G3-PEG6k-OH, Dendron functionalized PEG, generation 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

form

solid

mol wt

PEG average Mn 6,000
average Mn 8,000
generation 3

no. Surface Groups

16

mp

45-50 °C

functional group

hydroxyl

Looking for similar products? Visit Product Comparison Guide

Other Notes

average PEG Mn = 6,000; esterified with 16 OH groups

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qigang Wang et al.
Nature, 463(7279), 339-343 (2010-01-22)
With the world's focus on reducing our dependency on fossil-fuel energy, the scientific community can investigate new plastic materials that are much less dependent on petroleum than are conventional plastics. Given increasing environmental issues, the idea of replacing plastics with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service