Skip to Content
Merck
All Photos(3)

Key Documents

750085

Sigma-Aldrich

5,5′-Bis(trimethylstannyl)-2,2′-bithiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22S2Sn2
CAS Number:
Molecular Weight:
491.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

96-100 °C

SMILES string

C[Sn](C)(C)c1ccc(s1)-c2ccc(s2)[Sn](C)(C)C

InChI

1S/C8H4S2.6CH3.2Sn/c1-3-7(9-5-1)8-4-2-6-10-8;;;;;;;;/h1-4H;6*1H3;;

InChI key

DOIRPCDOGSNNCS-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service