Skip to Content
Merck
All Photos(1)

Key Documents

749796

Sigma-Aldrich

Indium tin oxide coated PET

surface resistivity 300 Ω/sq, L × W × thickness 1 ft × 1 ft × 5 mil, sheet

Synonym(s):

ITO-PET, Polyethylene terephthalate film, ITO coated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

sheet

surface resistivity

300 Ω/sq

L × W × thickness

1 ft × 1 ft × 5 mil

SMILES string

O=[In]O[In]=O.O=[Sn]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide coated polyethylene terephthalate (PET) film is optically transparent and electrically conductive.The ITO coating is protected by a thin, transparent film, that can be easily peeled off.

Application

ITO-PET may be used in fabricating flexible OLEDs. ITO/PET may be used as a reference anode in conventional flexible polymer based organic solar cells (OSCs).

Features and Benefits

The polyester film products comprise sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of the glass substrates.
These polyester film products comprise of sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of glass substrates.

Physical properties

Thickness of ITO coating is 240 Å

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and Flexible ITO-Free Organic Solar Cells Using Highly Conductive Polymer Anodes.
Na SI, et al.
Advanced Materials, 20(21), 4061-4067 (2008)
Modular mass spectrometric tool for analysis of composition and phosphorylation of protein complexes
Blethrow JD, et al.
PLoS ONE, 2(4), e358-e358 (2007)
High-Performance Organic Light-Emitting Diodes Using ITO Anodes Grown on Plastic by Room-Temperature Ion-Assisted Deposition
Yang Y, et al.
Advanced Materials, 16(4), 321-324 (2004)

Articles

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service