Skip to Content
Merck
All Photos(1)

Documents

749257

Sigma-Aldrich

2,2′-Bis[4-(trifluoromethyl)phenyl]-5,5′-bithiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10F6N2S2
CAS Number:
Molecular Weight:
456.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder or crystals

mp

230-235 °C

semiconductor properties

N-type (mobility=1.83 cm2/V·s)

SMILES string

FC(F)(F)c1ccc(cc1)-c2ncc(s2)-c3cnc(s3)-c4ccc(cc4)C(F)(F)F

InChI

1S/C20H10F6N2S2/c21-19(22,23)13-5-1-11(2-6-13)17-27-9-15(29-17)16-10-28-18(30-16)12-3-7-14(8-4-12)20(24,25)26/h1-10H

InChI key

YPPDUFZJMJWOLJ-UHFFFAOYSA-N

Application

This material is used as an n-type semiconductor for OFET devices. It forms a closely packed 2-dimensional columnar structure leading to a high performance n-type FET.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinji Ando et al.
Journal of the American Chemical Society, 127(43), 14996-14997 (2005-10-27)
Novel thiazole oligomers and thiazole/thiophene co-oligomers with trifluoromethylphenyl groups were developed as n-type semiconductors for OFETs. They showed excellent n-type performances with high electron mobilities. A 5,5'-bithiazole with trifluoromethylphenyl groups forms a closely packed two-dimensional columnar structure leading to a

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service