Skip to Content
Merck
All Photos(1)

Key Documents

744379

Sigma-Aldrich

Trimethylamine

≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.09 (vs air)

vapor pressure

13.3 psi ( 21 °C)

Assay

≥99.5%

form

gas

autoignition temp.

374 °F

expl. lim.

11.6 %

bp

3-4 °C (lit.)

mp

−117 °C (lit.)

density

0.63 g/mL at 20 °C (lit.)

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylamine is a tertiary amine used as a versatile building block in organic synthesis to produce choline chloride, surfactants and solvents, rubber chemicals, tetramethylammonium hydroxide, cationic starches, phase transfer catalysts, ion-exchange resins, and some basic dyes.

Application

Trimethylamine can be used as:
  • A base in dehydrochlorinations, nucleophilic substitutions, 1,4-additions, and aldol-type condensation reactions.
  • A reactant for the preparation of trimethylamine-based dicationic ionic liquids for the extraction of phenolic compounds from oil mixtures.

Packaging

500ml high pressure aluminum cylinder equipped with a stainless steel valve according to DIN 477-1.

Other Notes

For 744379-280G-EU and 744379-280G packaging: 500 mL of aluminium cylinder equipped with stainless steel valve.

Recommended products

For 744379-280G-EU and 744379-280G: outlet fitting: DIN 477-1 no. 1, Z742162 or Z742164 are the recommended regulators.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylamines
van Gysel AB and Musin W
Ullmann's Encyclopedia of Industrial Chemistry, 1-16 (2000)
Analytical method for monitoring airborne trimethylamine using solid phase micro-extraction and gas chromatography-flame ionization detection
Chien Y-C, et al.
Analytica Chimica Acta, 419(1), 73-79 (2000)
Trimethylamine
Sorgi KL
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 39(6), 501-505 (2007)
Kin Man Au et al.
Science advances, 6(27), eaba8564-eaba8564 (2020-09-15)
Activation of the innate immune system and natural killer (NK) cells has been a key effort in cancer immunotherapy research. Here, we report a nanoparticle-based trispecific NK cell engager (nano-TriNKE) platform that can target epidermal growth factor receptor (EGFR)-overexpressing tumors
Highly efficient extraction of phenolic compounds from oil mixtures by trimethylamine-based dicationic ionic liquids via forming deep eutectic solvents
Ji Youan, et al.
Fuel Processing Technology, 171(6), 183-191 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service