Skip to Content
Merck
All Photos(1)

Key Documents

744255

Sigma-Aldrich

Sulfur dioxide

≥99.98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2
CAS Number:
Molecular Weight:
64.06
Beilstein:
3535237
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.26 (21 °C, vs air)

Quality Level

vapor pressure

1779 mmHg ( 21 °C)

Assay

≥99.98%

form

gas

bp

−10 °C (lit.)

mp

−73 °C (lit.)

density

1.25 g/mL at 25 °C (lit.)

SMILES string

O=S=O

InChI

1S/O2S/c1-3-2

InChI key

RAHZWNYVWXNFOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfur dioxide (SO2) can be used:
  • In the palladium catalyzed aminosulfonylation reaction.[1]
  • To prepare 1,4-diazabicyclo[2.2.2]octane [DABCO. (SO2)2] or DABSO, which is used as a reagent to synthesize N-aminosulfonamides via aminosulfonylation process using palladium catalyst.[1][2]
  • To synthesize polysulfobetaine (PSB/SO2) copolymer.[3]

Packaging

500ml high pressure aluminum cylinder equipped with a stainless steel valve according to DIN 477-1.

Other Notes

For 744255-450G-EU packaging: 500 mL of aluminium cylinder equipped with brass valve.

Recommended products

For 744255-450G-EU: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742161 are the recommended regulators.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and cyclopolymerization of diallylammoniomethanesulfonate
Ali, Shaikh A and Al-Hamouz OCS
Polym. Eng. Sci., 53(11), 2378-2388 (2013)
Palladium-catalysed aminosulfonylation of aryl-, alkenyl-and heteroaryl halides: scope of the three-component synthesis of N-aminosulfonamides
Emmett EJ, et al.
Organic & Biomolecular Chemistry, 10(20), 4007-4014 (2012)
DABCO-bis (sulfur dioxide), DABSO, as a convenient source of sulfur dioxide for organic synthesis: utility in sulfonamide and sulfamide preparation.
Woolven H, et al.
Organic Letters, 13(18), 4876-4878 (2011)
Sulfur dioxide.
R Von Burg
Journal of applied toxicology : JAT, 16(4), 365-371 (1996-07-01)
J Neil Cape et al.
Environment international, 29(2-3), 201-211 (2003-04-05)
The regulation of the emissions of 'traditional' primary air pollutants (fluorides, sulfur dioxide) has changed the pattern of exposure of ecological systems, with greatly reduced exposure close to sources, but with a smaller effect in some remote areas. Measurements show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service