Skip to Content
Merck
All Photos(1)

Key Documents

74115

Sigma-Aldrich

trans-4-Nitrocinnamaldehyde

technical, ≥97.0% (T)

Synonym(s):

trans-3-(4-Nitrophenyl)-2-propenal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO3
CAS Number:
Molecular Weight:
177.16
Beilstein:
1565424
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥97.0% (T)

form

crystals

mp

139-142 °C

functional group

aldehyde
nitro

SMILES string

[O-][N+](=O)c1ccc(\C=C\C=O)cc1

InChI

1S/C9H7NO3/c11-7-1-2-8-3-5-9(6-4-8)10(12)13/h1-7H/b2-1+

InChI key

ALGQVMMYDWQDEC-OWOJBTEDSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Eder et al.
Mutagenesis, 6(4), 261-269 (1991-07-01)
Seventeen cinnamaldehydes, cinnamic acids, 2-furylacroleins and related compounds were tested in the Salmonella preincubation reversion assay and in the SOS chromotest. Of eight compounds containing nitrogroups, seven were clearly mutagenic in the presence of S9 mix and six in its
I Baburina et al.
Biochemistry, 37(5), 1245-1255 (1998-03-07)
The residue C221 on pyruvate decarboxylase (EC. 4.1.1.1) from Saccharomyces cerevisiae has been shown to be the site where the substrate activation cascade is triggered [Baburina et al. (1994) Biochemistry 33, 5630-5635] and is located on the beta domain [Arjunan

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service